Learn More
Masterflex™ Threaded Barb Adapter Kits, PVDF
Product Code. 11770429
Click to view available options
:
1/Pk
Product Code. 11770429
Brand: Masterflex™ 3151436
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Organize the fittings at your workbench with these convenient fittings kits
  • Designed for quick and easy assembly without tools.
  • Combine these simple and economical fittings with flexible tubing.
  • For use in low-pressure applications (generally less than 30psi).
Specifications
Description Barb x NPT(M), Large
Material PVDF
Packaging Quantity 1/Pk.
Product Type Threaded Barb Adapter Kit