Learn More
BOLA Thread Adaptor GL
Description
- Straight transition fitting made of PTFE, two GL-threaded connections.
- Universal chemical resistance, the medium is only exposed to PTFE.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Autoclavable | Autoclavable |
| Thread Size | GL 14 |
| Color | White |