Learn More
Thiourea-13C,15N2, TRC
Specifications
Specifications
| Chemical Name or Material | Thiourea-13C,15N2 |
| CAS | 285977-83-5 |
| Molecular Formula | 13C H4 15N2 S |
| Quantity | 50 mg |
| Synonym | Thio-pseudourea-13C,15N2, β-Thiopseudourea-13C,15N2, Thiocarbamide-13C,15N2 |
| InChI Formula | InChI=1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)/i1+1,2+1,3+1 |
| SMILES | [15NH2][13C](=S)[15NH2] |
| IUPAC Name | bis(^{15}N)(azanyl)(1^{13}C)methanethione |
| Molecular Weight (g/mol) | 79.1 |
| Formula Weight | 79.0069 |