missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2-boronic acid, 97%

Product Code. p-7026825
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446266 1 g 1g
11456266 5 g 5g
11466266 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446266 Supplier Thermo Scientific Alfa Aesar Supplier No. B23071.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6165-68-0 | C4H5BO2S | 127.95 g/mol

Thiophene-2-boronic acid is a reagent used for Palladium-catalyzed Suzuki-Miyaura cross-couplings, Copper-catalyzed nitration reactions, Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer, Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters. It is also used in the preparation of Amino pyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2-boronic acid is a reagent used for Palladium-catalyzed Suzuki-Miyaura cross-couplings, Copper-catalyzed nitration reactions, Chain-growth catalyst transfer polycondensation of conjugated alternating copolymer, Ferric perchlorate-promoted reaction of fullerene to give fullerenyl boronic esters. It is also used in the preparation of Amino pyridine-based inhibitors of mitotic kinase Nek2 with potential antipoliferative effects in cancer tumors.

Solubility
Soluble in methanol.

Notes
Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6165-68-0
Molecular Formula C4H5BO2S
Molecular Weight (g/mol) 127.95
MDL Number MFCD00151850
InChI Key ARYHTUPFQTUBBG-UHFFFAOYSA-N
Synonym thiophene-2-boronic acid, 2-thiopheneboronic acid, 2-thienylboronic acid, thienylboronic acid, 2-thiopheneboronicacid, thiophen-2-ylboranediol, thiophene2-boronic acid, thiophen-2-yl boronic acid, 2-thiophenylboric acid, boronic acid, thienyl
PubChem CID 2733960
IUPAC Name thiophen-2-ylboronic acid
SMILES OB(O)C1=CC=CS1

Specifications

Melting Point 128°C to 131°C
Quantity 1 g
Beilstein 112375
Solubility Information Soluble in methanol.
Formula Weight 127.96
Percent Purity 98%
Chemical Name or Material Thiophene-2-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.