missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thioglycollate Medium USP

Product Code. 12697198
Change view
Click to view available options
Fill Weight:
100 to 101.5 g
20 to 20.3 g
9 ± 0.5mL
Format:
125 mL Sirop Bottle
Tube
Universal Bottle
Unit Size:
Each
Pack of 10
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Fill Weight Format unitSize
12697198 20 to 20.3 g Universal Bottle Each
13295789 9 ± 0.5mL Tube Pack of 50
13265439 100 to 101.5 g 125 mL Sirop Bottle Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12697198 Supplier Thermo Scientific™ Supplier No. BO0211G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cultivate aerobic and anaerobic organisms in the performance of sterility tests with the convenience of prepared media tubes.

Cultivate aerobic and anaerobic organisms in the performance of sterility tests with Thermo Scientific™ Thioglycollate Medium USP prepared media tubes. The medium is prepared according to the formula specified in the US Pharmacopoeia1 for the performance of sterility tests, and conforms to formulations detailed in the British and European Pharmacopoeia 2,3.

Thioglycollate Medium USP is suitable for the cultivation of both aerobic and anaerobic organisms. No paraffin or special seal is necessary nor is an anaerobic jar required for the cultivation of anaerobes. It is well buffered, so that acid or alkaline inocula produce negligible alteration in the reaction of the medium. The sodium thioglycollate content of the medium will neutralize the bacteriostatic effect of mercurial compounds used as preservatives in solutions for injection, etc. If the solution undergoing test contains a bacteriostatic substance, it is necessary, in order to avoid a false negative result, to establish the bacteriostatic activity of the product by the method described in the US Pharmacopoeia1. Thioglycollate Medium USP is also recommended for the cultivation of Clostridium species.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw/yellow with or without red layer
Content And Storage 2° to 25°C
Format Universal Bottle
Manufacturing Origin United Kingdom
Product Type Microbiology Media
Fill Weight 20 to 20.3 g
Packaging Type 24 universal bottles with screw cap closures in a box
Quantity 24/Pk.

For In Vitro Diagnostic Use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.