Learn More
Thieno[3,2-c]pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | Thieno[3,2-c]pyridine |
| CAS | 272-14-0 |
| Molecular Formula | C7 H5 N S |
| Quantity | 250 mg |
| Synonym | Thieno[3,2-c]pyridine, Ticlopidine Hydrochloride Imp. A (EP) |
| InChI Formula | InChI=1S/C7H5NS/c1-3-8-5-6-2-4-9-7(1)6/h1-5H |
| SMILES | c1cc2sccc2cn1 |
| IUPAC Name | thieno[3,2-c]pyridine |
| Molecular Weight (g/mol) | 135.19 |
| Formula Weight | 135.0143 |