Learn More
Thiazovivin, MedChemExpress
Product Code. 16488287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488287 5 mg 5mg
16466136 100 mg 100mg
16415598 10 mg 10mg
16401767 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16488287 Supplier Medchem Express Supplier No. HY13257
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Thiazovivin is a potent ROCK inhibitor, which can protect human embryonic stem cells. Thiazovivin improves the efficiency of iPSC generation.
Thiazovivin is a potent ROCK inhibitor, which can protect human embryonic stem cells. Thiazovivin improves the efficiency of iPSC generation.
Specifications
Chemical Name or Material Thiazovivin
Physical Form Solid
CAS 1226056-71-8
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C15H13N5OS
For Use With (Application) Cancer-Kinase/protease
Solubility Information DMSO : ≥ 30 mg/mL (96.35 mM)
SMILES O=C(C1=CSC(NC2=NC=NC=C2)=N1)NCC3=CC=CC=C3
Molecular Weight (g/mol) 311.36
Formula Weight 311.36
Percent Purity 99.31%
Grade Research