missing translation for 'onlineSavingsMsg'
Learn More

Thiamphenicol, MedChemExpress

Product Code. p-200099144
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16428336

Brand: Medchem Express HYB0479

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Thiamphenicol (Thiophenicol), a methyl-sulfonyl derivative of Chloramphenicol, is a broad-spectrum antimicrobial antibiotic. Thiamphenicol acts by binding to the 50S ribosomal subunit, leading to inhibition of protein synthesis and bacteriostatic effect (against Gram-negative, Gram-positive aerobic and anaerobic bacteria).

Thiamphenicol (Thiophenicol), a methyl-sulfonyl derivative of Chloramphenicol, is a broad-spectrum antimicrobial antibiotic. Thiamphenicol acts by binding to the 50S ribosomal subunit, leading to inhibition of protein synthesis and bacteriostatic effect (against Gram-negative, Gram-positive aerobic and anaerobic bacteria).

Specifications

Chemical Name or Material Thiamphenicol
Physical Form Solid
CAS 15318-45-3
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H15Cl2NO5S
For Use With (Application) COVID-19-immunoregulation
Synonym Thiophenicol Dextrosulphenidol
Solubility Information DMSO : 100 mg/mL (280.73 mM; Need ultrasonic) ∣H2O : 2 mg/mL (5.61 mM; Need ultrasonic)
SMILES O=C(N[C@H](CO)[C@H](O)C1=CC=C(S(=O)(C)=O)C=C1)C(Cl)Cl
Molecular Weight (g/mol) 356.22
Formula Weight 356.22
Percent Purity 99.38%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.