Learn More
Thiabendazole, TRC
Product Code. 30138670
Change view
Click to view available options
Quantity:
50 mg
1 g
100 g
Unit Size:
100g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138670 1 g 1g
30138227 100 g 100g
17922095 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138670 Supplier Toronto Research Chemicals Supplier No. TRCT3441501G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 148-79-8
Molecular Formula C10 H7 N3 S
Molecular Weight (g/mol) 201.25
Synonym 1H-Benzimidazole, 2-(4-thiazolyl)-, Benzimidazole, 2-(4-thiazolyl)- (6CI,8CI), 2-(4-Thiazolyl)-1H-benzimidazole, 2-(1,3-Thiazol-4-yl)-1H-1,3-benzodiazole, 2-(1,3-Thiazol-4-yl)-1H-benzimidazole, 2-(1,3-Thiazol-4-yl)benzimidazole, 2-(4-Thiazolyl)-1H-benzoimidazole, 2-(4-Thiazolyl)benzimidazole, 2-(4-Thiazoyl)benzimidazole, 2-(4-Triazolyl)benzimidazole, 2-(4'-Thiazolyl)benzimidazole, 2-(4'-Thiazoyl)benzimidazole, 4-(1H-Benzimidazol-2-yl)thiazole, 5-(4-Thiazolyl)benzimidazole, Amolden HS, Biogard TBH, Bovizole, Chemviron TK 100, Cropasal, Drawipas, Eprofil, Equizole, G 491, Hokkustar HP, Hokustar HP, Irgaguard F 3000, Lombristop, MK 360, MSD 18, Marukacide M 101, Mertect, Mertect 160, Mertect 340F, Mertect LSP, Metasol TK 100, Metasol TK 25, Metasol TK 25AD, Metasol TK 50AD, Mintesol, Mintezol, Minzolum, NSC 525040, NSC 90507, Nemapan, Neo-Sintol AF 75, Omnizole, Ormogal, Pitrizet, Polival, Sanaizol 100, Sanaizol TBZ-FL 25, Sistesan, Storite, Syntol M 100, TBZ, TBZ (fungicide), TBZ 6, TBZ 60W, TK 25AD, Tebuzate, Tecta, Tectab, Tecto, Tecto 10P, Tecto 40F, Tecto 60, Tecto B, Thiaben, Thiabendazol, Thiabendazole, Thiabendole, Thiabenzole, Thibendole, Thibenzol, Thibenzole, Thibenzole 200, Tiabenda, Tiabendazol, Tiabendazole, Tibimix 20, Top Form Wormer, Triasox
IUPAC Name 4-(1H-benzimidazol-2-yl)-1,3-thiazole
SMILES c1ccc2[nH]c(nc2c1)c3cscn3
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H7N3S/c1-2-4-8-7(3-1)12-10(13-8)9-5-14-6-11-9/h1-6H,(H,12,13)
Formula Weight 201.0361
Chemical Name or Material Thiabendazole