Learn More
Thiabendazole, MedChemExpress
Product Code. 16480848
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480848 5 g 5g
16402119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16480848 Supplier Medchem Express Supplier No. HYB0263
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Thiabendazole inhibites the mitochondrial helminth-specific enzyme, fumarate reductase, with anthelminthic property.
Thiabendazole inhibites the mitochondrial helminth-specific enzyme, fumarate reductase, with anthelminthic property.
Specifications
Chemical Name or Material Thiabendazole
Physical Form Solid
CAS 148-79-8
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C10H7N3S
For Use With (Application) COVID-19-immunoregulation
Synonym 2-(4-Thiazolyl)benzimidazole
Solubility Information DMSO : 50 mg/mL (248.45 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C1(C2=CSC=N2)=NC3=CC=CC=C3N1
Molecular Weight (g/mol) 201.25
Formula Weight 201.25
Percent Purity 98.0%
Grade Research