Learn More
Sonoco™ ThermoSafe PolarPack™ Foam Bricks
Product Code. 13079342
Change view
Click to view available options
Dimensions (LxWxH):
16.51 x 11.43 x 3.81 cm
20.9 x 20.3 x 1.9 cm
22.8 x 12.1 x 1.9 cm
Weight (English):
17 oz.
18 oz.
24 oz.
Weight (Metric):
482 g
0.68 kg
0.72 kg
Unit Size:
Pack of 24
Pack of 28
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (LxWxH) Weight (English) Weight (Metric) Unit Size
13079342 16.51 x 11.43 x 3.81 cm 24 oz. 0.68 kg Pack of 24
17879271 22.8 x 12.1 x 1.9 cm 17 oz. 482 g Pack of 24
13059342 20.9 x 20.3 x 1.9 cm 18 oz. 0.72 kg Pack of 28
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13079342 Supplier Sonoco™ Supplier No. FPP24
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
Refrigerant bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.
  • Provide refrigeration for transport of food, pharmaceuticals, and medical products.
  • Gel will phase change at 0° C.
  • PolarPack Foam Bricks maintain designated packing configurations throughout the trip.
  • For frozen or refrigerated shipments.
  • Alternative to wet ice.
  • Available in several sizes
  • TRUSTED_SUSTAINABILITY
    Specifications
    Quantity 24/Cs.
    Width (Metric) Exterior 11.43 cm
    Length (English) Exterior 6.5 in.
    Width (English) Exterior 4.5 in.
    Length (Metric) Exterior 16.51 cm
    Height (English) Exterior 1.5 in.
    Type Refrigerant Ice Pack
    Height (Metric) Exterior 3.81 cm
    Weight (English) 24 oz.
    Weight (Metric) 0.68 kg
    Dimensions (LxWxH) 16.51 x 11.43 x 3.81 cm
    Freezing Point 0°C (32°F)