Learn More
GFL Gesellschaft Fuer Labortec™ Thermolab™ 10770 Shaking Waterbath
Product Code. 10567801
Change view
Click to view available options
Weight (Metric):
28 / 36 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Weight (Metric) Unit Size
10567801 28 / 36 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10567801 Supplier GFL Gesellschaft Fuer Labortec™ Supplier No. 1070
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Shaking Motion Reciprocating
Temperature Range 5°C to 100°C
Type Shaking Water Bath
Oscillations per Minute 2 to 50 rpm
Power Consumption 1.3 kW
Speed 2 to 50rpm
Width (Metric) 625 mm
Width (Metric) Interior 175 mm
Height (Metric) 270 mm
Height (Metric) Interior 100 mm
Depth (Metric) 556 mm
Depth (Metric) Interior 175 mm
Weight (Metric) 28 / 36 kg
Electrical Requirements 230 V
Voltage 230 V
Frequency 50/60 Hz