Learn More
TFA Dostmann Thermo-Timer
Click to view available options
Product Type:
Timer
Unit Size:
Each
Description
The Thermo-Timer timer gives you the option of switching devices up to 3600 watts depending on the temperature or time or a combination of temperature and time. The waterproof cable sensor enables temperature measurement in remote environments and in liquids.
- For time and / or temperature controlled switching
- Timer mode (three possible on/off time pairs)
- Thermostat mode (automatic activation when falling below a certain temperature)
- Thermo-Timer-Mode (combination)
- Measurement error ± 1°C, 1 m cable
- Switching capacity up to 3500 watts (16 A at 230 V)
Specifications
Specifications
| Product Type | Timer |