Learn More
Epredia™ Shandon™ Tissue-Loc™ Tissue Cassettes IV
Shop All Thermo Scientific Products
Click to view available options
Color:
Blue
Lilac
White
Yellow
Unit Size:
Pack of 250
Description
- Disposable
- Holds tissue specimens during embedding, processing
- Pore style: Small slots
- Supplied in box of 250 cassettes with attached lids
Specifications
Specifications
| Pore Style | Small Slots |
| Closure Type | Hinged |
| Color | Lilac |