Learn More
Thermographic Measurements™ Thermax™ 5 Level Temperature Indicator Strips
Provide a fast, simple temperature monitoring system based on proprietary color change technology. TMC Hallcrest™ Thermax™ 5 Level Temperature Indicator Strips are irreversible and record the highest temperature achieved.
Brand:  Thermographic Measurements™ M05STHRGFD01P1
Product Code. 11710899
  • 9.55 € / Pack of 10
Only null left
Estimated Shipment: 19-06-2026
Add to basket
Description
Description
  • Heat-sensitive label to monitor temperature
  • Inexpensive and easy to use.
  • Immediate response.
  • Self-adhesive
  • Discreet
  • Irreversible
  • Oil, water and steam resistant.
  • Accuracy tolerances: (29-99°C (+/-1°C) / 100 -154°C (+/-1.5°C) 160-290°C (+/-1%+1°C).