missing translation for 'onlineSavingsMsg'
Learn More

Thalidomide-4-OH, MedChemExpress

Product Code. 16441556
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16441556

Brand: Medchem Express HY103596

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Thalidomide-4-OH (Cereblon ligand 2) is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide-4-OH (Cereblon ligand 2) can be connected to the ligand for protein by a linker to form PROTACs.

Thalidomide-4-OH (Cereblon ligand 2) is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide-4-OH (Cereblon ligand 2) can be connected to the ligand for protein by a linker to form PROTACs.

Specifications

Chemical Name or Material Thalidomide-4-OH
CAS 5054-59-1
Color Light Gray
Purity Grade Notes Research
Molecular Formula C13H10N2O5
Quantity 500 mg
Synonym Cereblon ligand 2 E3 ligase Ligand 2
Solubility Information DMSO : ≥ 36.5 mg/mL (133.10 mM)
SMILES OC1=CC=CC(C(N2C3C(NC(CC3)=O)=O)=O)=C1C2=O
Molecular Weight (g/mol) 274.23
Formula Weight 274.23
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.