Learn More
Thalidomide 4-fluoride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Thalidomide 4-fluoride (Cereblon ligand 4) is the Thalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Thalidomide 4-fluoride (Cereblon ligand 4) can be connected to the ligand for IRAK4 protein by a linker to form PROTAC IRAK4 degrader-1 (HY-129966).
Specifications
Specifications
| Chemical Name or Material | Thalidomide 4-fluoride |
| CAS | 835616-60-9 |
| Color | Purple |
| Purity Grade Notes | Research |
| Molecular Formula | C13H9FN2O4 |
| Quantity | 100 mg |
| Synonym | Cereblon ligand 4 E3 ligase Ligand 4 |
| Solubility Information | DMSO : 300 mg/mL (1086.09 mM; Need ultrasonic) |
| SMILES | O=C1N(C(CC2)C(NC2=O)=O)C(C3=C1C=CC=C3F)=O |
| Molecular Weight (g/mol) | 276.22 |
| Show More |