missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ THADA Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
THADA (thyroid adenoma associated), also known as GITA, is a 1,953 amino acid protein that is expressed in testis, thyroid, pancreas, stomach and small intestine. Chromosomal aberrations in the gene encoding THADA are associated with benign thyroid adenomas, suggesting a potential role for THADA in tumorigenesis. The THADA gene maps to human chromosome 2, which houses over 1,400 genes and comprises nearly 8% of the human genome. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the chromosome 2-localized ABCA12 gene, while the lipid metabolic disorder sitosterolemia is associated with defects in the ABCG5 and ABCG8 genes, which also map to chromosome 2.
Specifications
Specifications
| Antigen | THADA |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | THADA |
| Gene Accession No. | A8C756, Q6YHU6 |
| Gene Alias | 6530405K19; A930021O22; ARMC13; AU019899; BC052885; death receptor-interacting protein; gene inducing thyroid adenomas protein; GITA; KIAA1767; putative death receptor-interacting protein; RGD1560519; Thada; THADA armadillo repeat containing; THADA, armadillo repeat containing; thyroid adenoma associated; thyroid adenoma-associated protein; Thyroid adenoma-associated protein homolog |
| Gene Symbols | THADA |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?