missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TGF-beta 3 Antibody (TGFB3/4801), Novus Biologicals™
Shop All Bio Techne ProductsDescription
TGF-beta 3 Monoclonal antibody specifically detects TGF-beta 3 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | TGF-beta 3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | TGFB3/4801 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | ARVD, FLJ16571, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?