missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF-beta 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03276
This item is not returnable.
View return policy
Description
TGF-beta 1 Polyclonal antibody specifically detects TGF-beta 1 in Human, Mouse, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| TGF-beta 1 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, ELISA 1:10000 - 1:20000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 5 μg/mL, Immunohistochemistry-Frozen 1:10-1:500 | |
| P01137 | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Equine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 20mM Potassium Phosphate (pH 7.2) and 0.15M NaCl | |
| CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1 | |
| Synthetic peptide from human TGF-beta 1. | |
| 0.05 mg | |
| Angiogenesis | |
| 7040 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction