Learn More
Novus Biologicals™ TGF-beta 1 Antibody (Genzyme patent anti-TGF-beta), Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28452-100ug
 View more versions of this product
Product Code. 30044720
  • 338.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
Description
Description
TGF-beta 1 Monoclonal antibody specifically detects TGF-beta 1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TGF-beta 1
Monoclonal
Unconjugated
P01137
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Genzyme patent anti-TGF-beta
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CEDLAP, DPD1, latency-associated peptide, TGF beta, TGFB, TGFbeta, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
TGFb1
100 μg
Angiogenesis, Apoptosis, Asthma, Cancer, Immunology, Phospho Specific, Signal Transduction
7040