missing translation for 'onlineSavingsMsg'
Learn More

TG100-115, MedChemExpress

Product Code. 16413278
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16413278

Brand: Medchem Express HY1011110mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress TG100-115 is a selective PI3Kγ/PI3Kδ inhibitor with IC50s of 83 and 235 nM, respectively.

TG100-115 is a selective PI3Kγ/PI3Kδ inhibitor with IC50s of 83 and 235 nM, respectively.

Specifications

Chemical Name or Material TG100-115
CAS 677297-51-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H14N6O2
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (96.23 mM; ultrasonic and warming and adjust pH to 2 with HCl and heat to 60°C)
SMILES NC1=C(C(N=C2C3=CC(O)=CC=C3)=NC(N)=N1)N=C2C4=CC(O)=CC=C4
Molecular Weight (g/mol) 346.34
Formula Weight 346.34
Percent Purity 98.04%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.