Learn More
TFPT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16886253 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16886253 20 μL 20µL
16876253 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16886253 Supplier Proteintech Supplier No. 100972AP20UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
TFPT component of the chromatin-remodeling INO80 complex which, in addition to chromatin remodeling implicated in crucial functions such as DNA repair, checkpoint regulation, DNA replication, telomemere maintece and chromosome segregation. This protein appears to promote apoptosis in a TP53/p53-independent manner.
Specifications
Antigen TFPT
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TFPT
Gene Accession No. P0C1Z6, Q3U1J1, Q9JMG6
Gene Alias amida, FB1, INO80 complex subunit F, INO80F, Protein FB1, TCF3 fusion partner, TFPT
Gene Symbols Tfpt
Host Species Rabbit
Immunogen TFPT Fusion Protein Ag0131
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29844, 69714, 85423
Target Species Rat, Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG