Learn More
Tocris Bioscience™ TFC 007
GREENER_CHOICE
Potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor
Brand:  Tocris Bioscience™ 5108/10
Encompass
Product Code. 15716476
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
This item is not returnable. View return policy
Description
Description
TFC 007 is a potent hematopoietic prostaglandin D synthase (H-PGDS) inhibitor (IC50 = 83 nM). Inhibits PGD2 production and late phase nasal blockage in an animal model of allergic rhinitis. Exhibits negligible effects on COX-1, COX-2, 5-LOX, LTC4 synthase, TXA2 synthase and microsomal prostaglandin E2 synthase.
Specifications
Specifications
>98%