Learn More
Tocris Bioscience™ TFB-TBOA
Description
TFB-TBOA is a potent and selective glial glutamate transporter EAAT1 and EAAT2 inhibitor (IC50 values are 17, 22 and 300 nM for EAAT2, EAAT1 and EAAT3 respectively). Has no effect on EAAT4 and EAAT5, or a wide range of neuronal receptors and transporters. Attenuates glutamate-stimulated intracellular Na+ elevation in astrocytes in vitro (IC50 = 43 nM). Induces severe convulsions in vivo.
Specifications
Specifications
| Chemical Name or Material | (3S)-3-[[3-[[4-(Trifluoromethyl)benzoyl]amino]phenyl]methoxy]-L-aspartic acid |
| CAS | 480439-73-4 |
| Assay Percent Range | >98% |
| Quantity | 1 mg |
| Solubility | Soluble to 100mM in DMSO |
| Molecular Formula | C19H17F3N2O6 |
| Formula Weight | Observed MW: 426.35 |