missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TEX11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570190
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 77%; Dog: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
This gene is X-linked and is expressed in only male germ cells. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.
Specifications
| TEX11 | |
| Polyclonal | |
| Unconjugated | |
| TEX11 | |
| 4930565P14Rik; BB024231; Protein ZIP4 homolog; RGD1560239; SPGFX2; testis expressed 11; testis expressed gene 11; testis expressed sequence 11; testis-expressed protein 11; testis-expressed sequence 11 protein; testis-expressed sequence 11 protein; testis-expressed protein 11; Tex11; TGC1; TSGA3; ZIP4H | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 501588 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| 0 | |
| TEX11 | |
| synthetic peptide directed towards the N-terminal region of rat TEX11. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction