missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Tetramethylrhodamine (TAMRA) Azide (Tetramethylrhodamine 5-Carboxamido-(6-Azidohexanyl)), 5-isomer

Product Code. 10738225
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 10738225

Brand: Molecular Probes™ T10182

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Tetramethylrhodamine (TAMRA) Azide (Tetramethylrhodamine 5-Carboxamido-(6-Azidohexanyl)), 5-isomer

The red-fluorescent tetramethylrhodamine (TAMRA) azide can be reacted with terminal alkynes via a copper-catalyzed click reaction. Click chemistry describes a class of chemical reactions that use bio-orthogonal or biologically unique moities to label and detect a molecule of interest using a two-step procedure. The two-step reaction procedure involves a copper-catalyzed triazole formation of an azide and an alkyne. Click reactions have several characteristics: the reaction between the detection moieties is efficient; no extreme temperatures or solvents are required; the reaction product is stable; the components of the reaction are bioinert; and perhaps most importantly, no side reactions occur – the label and detection tags react selectively and specifically with one another. Unlike traditional chemical reactions utilizing succinimidyl esters or maleimides that target amines and sulfhydryls – functional groups that are not unique – click chemistry-labeled molecules can be applied to complex biological samples and be detected with unprecedented sensitivity due to extremely low background.
TRUSTED_SUSTAINABILITY

Specifications

Format Solid
Quantity 0.5 mg
Product Type Tetramethylrhodamine Azide
Molecular Weight (g/mol) 554.65 Da
Solubility DMSO (Dimethylsulfoxide)
Content And Storage Store at ≤-20°C, desiccated and protected from light.
Chemical Reactivity Alkyne
Detection Method Fluorescence
Reactive Group Azide
Shipping Condition Room Temperature
Reactive Moiety Amine, Azide
Label or Dye TAMRA™ Isomers, TMR (Tetramethylrhodamine)
Label Type Classic Dyes
Color Orange
Product Line Molecular Probes
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.