Learn More
Tetramethyl-d12-ammonium Bromide, CDN
Specifications
Specifications
| Chemical Name or Material | Tetramethylammonium-D12 Bromide |
| CAS | 284474-82-4 |
| Molecular Formula | C4 D12 N . Br |
| Quantity | 0.5 g |
| Synonym | Methan-d3-aminium, N,N,N-tri(methyl-d3)-, bromide (9 CI), Tetra(methyl-d3)ammonium bromide, Tetramethylammonium bromide-d12, Tetramethylammonium-d12 bromide, Tetramethylammonium-D12 bromide |
| InChI Formula | InChI=1 S/C4H12N.BrH/c1-5(2,3)4;/h1-4H3;1 H/q+1;/p-1/i1D3,2D3,3D3,4D3; |
| SMILES | [Br-].[2 H]C([2 H])([2 H])[N+](C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tetrakis(trideuteriomethyl)azanium;bromide |
| Molecular Weight (g/mol) | 166.123 |
| Formula Weight | 165.091 g/mol |
| Show More |