Learn More
Tetramethyl Bisphenol A-d6, TRC
Product Code. 30411802
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411802 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30411802 Supplier Toronto Research Chemicals Supplier No. TRCT30093225MG
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetramethyl Bisphenol A-d6
CAS 203578-30-7
Molecular Formula C19 D6 H18 O2
Quantity 25 mg
Synonym 2,2-Bis[3,5-(dimethyl-d6)-4-hydroxyphenyl]propane, Bisxylenol A-d6, NSC 73730-d6, 2,2-Bis[4-hydroxy-3,5-(dimethyl-d6)phenyl]propane, Tetramethylbisphenol A-d6, 3,3',5,5'-Tetramethyl-4,4'-dihydroxydiphenyl-2,2-propane-d6, 4,4'-Isopropylidenebis[2,6-(dimethyl-d6)phenol], 4,4'-Isopropylidenedi-2,6-xylenol-d6
InChI Formula InChI=1S/C19H24O2/c1-11-7-15(8-12(2)17(11)20)19(5,6)16-9-13(3)18(21)14(4)10-16/h7-10,20-21H,1-6H3/i5D3,6D3
SMILES [2H]C([2H])([2H])C(c1cc(C)c(O)c(C)c1)(c2cc(C)c(O)c(C)c2)C([2H])([2H])[2H]
IUPAC Name 4-[1,1,1,3,3,3-hexadeuterio-2-(4-hydroxy-3,5-dimethylphenyl)propan-2-yl]-2,6-dimethylphenol
Molecular Weight (g/mol) 290.43
Formula Weight 290.215