Learn More
Tetrahydrozoline Hydrochloride, TRC
Product Code. 30135752
Change view
Click to view available options
Quantity:
50 mg
1 g
2.5 g
Unit Size:
1g
2.5g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135752 50 mg 50mg
17953985 2.5 g 2.5g
17922765 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135752 Supplier Toronto Research Chemicals Supplier No. TRCT29610050MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 522-48-5
Molecular Formula C13 H16 N2 . Cl H
Molecular Weight (g/mol) 236.74
Synonym Tetryzoline Hydrochloride, 4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole hydrochloride (1:1), 4,5-Dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-1H-imidazole monohydrochloride, 1H-Imidazole, 4,5-dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-, hydrochloride (1:1) (ACI), 1H-Imidazole, 4,5-dihydro-2-(1,2,3,4-tetrahydro-1-naphthalenyl)-, monohydrochloride (9CI), 2-Imidazoline, 2-(1,2,3,4-tetrahydro-1-naphthyl)-, monohydrochloride (7CI, 8CI), 2-(1,2,3,4-Tetrahydro-1-naphthyl)-2-imidazoline hydrochloride, 2-(1,2,3,4-Tetrahydronaphthalen-1-yl)-4,5-dihydro-1H-imidazole hydrochloride, Murine Plus, Soothe, Tetrahydrozoline hydrochloride, Tetryzoline hydrochloride, Tinarhinin, Tizine, Tyzanol, Tyzanol hydrochloride, Tyzine, Tyzine hydrochloride, Visine, Visine hydrochloride, Vislin, Yxin
IUPAC Name 2-(1,2,3,4-tetrahydronaphthalen-1-yl)-4,5-dihydro-1H-imidazole;hydrochloride
SMILES Cl.C1CC(C2=NCCN2)c3ccccc3C1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C13H16N2.ClH/c1-2-6-11-10(4-1)5-3-7-12(11)13-14-8-9-15-13;/h1-2,4,6,12H,3,5,7-9H2,(H,14,15);1H
Formula Weight 236.108
Percent Purity >95%
Chemical Name or Material Tetryzoline hydrochloride
Grade HPLC