missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydropyran-2-ylmethylamine, 97%, Thermo Scientific™

Product Code. 10735494
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10735494 1 g Amber Glass Bottle 1g
10799405 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10735494 Supplier Thermo Scientific Maybridge Supplier No. CC41213DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 683233-12-7
Molecular Formula C6H14ClNO
Molecular Weight (g/mol) 151.634
MDL Number MFCD06738971
InChI Key NGHRATGQJXKQIQ-UHFFFAOYSA-N
Synonym oxan-2-ylmethanamine hydrochloride, tetrahydropyran-2-ylmethylamine hydrochloride, c-tetrahydro-pyran-2-yl-methylamine hydrochloride, oxan-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-yl methanamine hydrochloride, tetrahydro-2h-pyran-2-ylmethyl amine hydrochloride, tetrahydro-2h-pyran-2-yl methyl amine hydrochloride, 2-aminomethyltetrahydropyran hydrochloride, tetrahydro-pyran-2-ylmethylamine hcl, 1-oxan-2-yl methanamine hydrochloride
PubChem CID 43811037
IUPAC Name oxan-2-ylmethanamine;hydrochloride
SMILES C1CCOC(C1)CN.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 151.64
Percent Purity 95%
Chemical Name or Material Tetrahydropyran-2-ylmethylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.