missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrocannabinol Antibody (THC3), PerCP, Novus Biologicals™

Product Code. 18662789 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18662789 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18662789 Supplier Novus Biologicals Supplier No. NBP307897PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Tetrahydrocannabinol Monoclonal antibody specifically detects Tetrahydrocannabinol in Nonspecies specific samples. It is validated for ELISA, Immunoassay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tetrahydrocannabinol
Applications ELISA, Immunoassay
Classification Monoclonal
Clone THC3
Conjugate PerCP
Formulation PBS
Gene Alias C21H30O2;Delta9-Tetrahydrocannabinol;Delta9-THC;Deltanyne;Dronabinol;Marinol;Tetrahydrocannabinol
Host Species Mouse
Immunogen Tetrahydrocannabinol (THC)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.