Learn More
Tetrahydro Furosemide, TRC
Product Code. 30425837
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30425837 Supplier Toronto Research Chemicals Supplier No. TRCT2935401G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Tetrahydro Furosemide
CAS 4793-38-8
Purity Grade Notes HPLC
Molecular Formula C12 H15 Cl N2 O5 S
Quantity 1 g
Synonym 5-(Aminosulfonyl)-4-chloro-2-[[(tetrahydro-2-furanyl)methyl]amino]benzoic acid, Anthranilic acid','4-chloro-5-sulfamoyl-N-(tetrahydrofurfuryl)- (7CI,8CI), Furosemide Imp. F (EP)
InChI Formula InChI=1S/C12H15ClN2O5S/c13-9-5-10(15-6-7-2-1-3-20-7)8(12(16)17)4-11(9)21(14,18)19/h4-5,7,15H,1-3,6H2,(H,16,17)(H2,14,18,19)
SMILES NS(=O)(=O)c1cc(C(=O)O)c(NCC2CCCO2)cc1Cl
IUPAC Name 4-chloro-2-(oxolan-2-ylmethylamino)-5-sulfamoylbenzoic acid
Molecular Weight (g/mol) 334.78
Formula Weight 334.039
Percent Purity >95