Learn More
Tetraethoxy-d20-silane, CDN
Specifications
Specifications
| Chemical Name or Material | Tetraethoxysilane-D20 |
| CAS | 272778-10-6 |
| Molecular Formula | C8 D19 H O4 Si |
| Quantity | 0.25 g |
| Synonym | Silicic acid (H4SiO4), tetra(ethyl-d5) ester (9 CI), Ethyl orthosilicate-d20, Silicon tetraethoxide-d20, Tetraethoxysilane-d20, Tetraethoxysilane-D20, Tetra(ethyl-d5) orthosilicate |
| InChI Formula | InChI=1 S/C8H20O4Si/c1-5-9-13(10-6-2,11-7-3)12-8-4/h5-8H2,1-4H3/i1D2,2D3,3D3,4D3,5D2,6D2,7D2,8D2 |
| SMILES | [2 H]C([2 H])C([2 H])([2 H])O[Si](OC([2 H])([2 H])C([2 H])([2 H])[2 H])(OC([2 H])([2 H])C([2 H])([2 H])[2 H])OC([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | tris(1,1,2,2,2-pentadeuterioethyl) 1,1,2,2-tetradeuterioethyl silicate |
| Molecular Weight (g/mol) | 227.445 |
| Formula Weight | 227.232 g/mol |
| Show More |