Learn More
Tetradecyl Pyridinium Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Tetradecylpyridinium Chloride |
| CAS | 2785-54-8 |
| Molecular Formula | C19 H34 N . Cl |
| Quantity | 500 mg |
| Synonym | Pyridinium','1-tetradecyl-','chloride (1:1), 1-Tetradecylpyridinium chloride (6CI,7CI), Pyridinium','1-tetradecyl-','chloride (8CI,9CI), 1-n-Tetradecanepyridinium chloride, Myristylpyridinium chloride, N-Tetradecylpyridinium chloride, TPC, TPC (onium compound |
| InChI Formula | InChI=1S/C19H34N.ClH/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-20-18-15-13-16-19-20;/h13,15-16,18-19H,2-12,14,17H2,1H3;1H/q+1;/p-1 |
| SMILES | [Cl-].CCCCCCCCCCCCCC[n+]1ccccc1 |
| IUPAC Name | 1-tetradecylpyridin-1-ium;chloride |
| Molecular Weight (g/mol) | 311.933 |
| Formula Weight | 311.238 |