missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) hydroxide solution, Pt 8-11% w/w (cont. Pt)

Product Code. p-7058638
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11346169

Brand: Thermo Scientific Alfa Aesar 042918.NH

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38201-97-7 | H14N4O2Pt | 297.22 g/mol

Tetraammineplatinum(II) hydroxide solution acts as a precursor for a platinum catalyst. It acts as noble metal catalyst which is primarily useful for homogeneous catalysis applications. It is used as a catalyst in hydrogenation, carbonylation, hydroformylation, chiral catalysis reactions and ruthenium dyes. Furthermore, it is used for C-X coupling reaction like Heck, Suzuki and Stille reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38201-97-7
Molecular Formula H14N4O2Pt
Molecular Weight (g/mol) 297.22
MDL Number MFCD00050044
InChI Key VSKCDODNDPOZKS-UHFFFAOYSA-L
IUPAC Name platinum(2+) tetraamine dihydroxide
SMILES N.N.N.N.[OH-].[OH-].[Pt++]

Specifications

Color Colorless
Odor Odorless
Linear Formula (NH3)4Pt(OH)2
Quantity 1 g
Solubility Information Miscible with water.
Formula Weight 297.23
Concentration or Composition (by Analyte or Components) Pt 8 to 11% w/w (cont. Pt)
Physical Form Liquid
Chemical Name or Material Tetraammineplatinum (II) hydroxide solution

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.