missing translation for 'onlineSavingsMsg'
Learn More

Tetraamminepalladium(II) chloride monohydrate, 99.9% (metals basis), Pd 39% min

Product Code. p-7048616
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11331668 2 g 2g
11341668 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331668 Supplier Thermo Scientific Alfa Aesar Supplier No. 011036.04
Add to basket
This item is not returnable. View return policy

CAS: 13933-31-8 | Cl2H14N4OPd | 263.46 g/mol

Tetraamminepalladium(II) chloride monohydrate is used to prepare trans-diamminedichloropalladium(II). It is also utilized in the preparation of mesoporous carbon supporting palladium nanoparticles by using graphite oxide. It is used as catalysts, reagents, and analytical reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13933-31-8
Molecular Formula Cl2H14N4OPd
Molecular Weight (g/mol) 263.46
MDL Number MFCD00151033
InChI Key WVCXSPJPERKPJS-UHFFFAOYSA-L
IUPAC Name palladium(2+) tetraamine hydrate dichloride
SMILES N.N.N.N.O.[Cl-].[Cl-].[Pd++]

Specifications

Melting Point 120°C (decomposition)
Color Colorless to Yellow
Density 1.91 g/mL
Quantity 2 g
UN Number UN3077
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 263.45 (245.43 Anhydrous)
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) Pd 39% min.
Physical Form Crystalline
Assay (metals basis)
Chemical Name or Material Tetraamminepalladium(II) chloride monohydrate
Show More Show Less
compliance-icons
Product Identifier
  • Tetraamminepalladium(II) chloride monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tetraamminepalladium(II) chloride monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.