Learn More
Tetra-O-pivaloyl-Alpha-D-glucopyranosyl Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | Tetra-O-pivaloyl-alpha-D-glucopyranosyl Bromide |
| CAS | 81058-27-7 |
| Molecular Formula | C26H43BrO9 |
| Quantity | 10 g |
| Synonym | 2,3,4,6-Tetrakis(2,2-dimethylpropanoate) alpha-D-Glucopyranosyl Bromide, 2,3,4,6-Tetra-O-pivaloyl-alpha-D-glucopyranosyl bromide |
| InChI Formula | InChI=1S/C26H43BrO9/c1-23(2,3)19(28)32-13-14-15(34-20(29)24(4,5)6)16(35-21(30)25(7,8)9)17(18(27)33-14)36-22(31)26(10,11)12/h14-18H,13H2,1-12H3/t14-,15-,16+,17-,18+/m1/s1 |
| SMILES | CC(C)(C)C(=O)OC[C@H]1O[C@H](Br)[C@H](OC(=O)C(C)(C)C)[C@@H](OC(=O)C(C)(C)C)[C@@H]1OC(=O)C(C)(C)C |
| IUPAC Name | [(2R,3R,4S,5R,6R)-6-bromo-3,4,5-tris(2,2-dimethylpropanoyloxy)oxan-2-yl]methyl 2,2-dimethylpropanoate |
| Molecular Weight (g/mol) | 579.52 |
| Formula Weight | 578.209 |