missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TET2 Polyclonal antibody specifically detects TET2 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | TET2 |
| Applications | ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 669 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | FLJ20032, MGC125715, tet methylcytosine dioxygenase 2 |
| Host Species | Rabbit |
| Immunogen | TET2 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human TET2. The immunogen is located within amino acids 20 - 70 of TET2. |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?