Learn More
TET1 Antibody (1462), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-15135
Product Code. 18077705
  • 680.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
TET1 Monoclonal antibody specifically detects TET1 in Human, Mouse, Rat samples. It is validated for Western Blot, ChIP assay, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Knockdown.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TET1
Monoclonal
1mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Knockdown Validated
TET1
Recombinant protein encompassing a sequence within the center region of human TET1. The exact sequence is proprietary.
Protein G purified
RUO
80312
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
IgG2b
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
1462
Unconjugated
bA119F7.1, CXXC finger 6, CXXC6, CXXC6FLJ10839, FLJ41442, KIAA1676CXXC-type zinc finger protein 6, LCX, LCXEC 1.14.11.n2, Leukemia-associated protein with a CXXC domain, methylcytosine dioxygenase TET1, Ten-eleven translocation 1 gene protein, ten-eleven translocation-1, tet oncogene 1
Mouse
235 kDa
0.1 mL
Primary
Human, Mouse, Rat