missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Enanthate-d3, TRC
Specifications
Specifications
| Chemical Name or Material | Testosterone Enanthate-d3 |
| Molecular Formula | C26H37D3O3 |
| Quantity | 10 mg |
| Synonym | 17β-Enanthoxyandrost-4-en-3-one-d3, 17β-Hydroxyandrost-4-en-3-one Enanthate-d3, 4-Androsten-3-one 17β-enanthate-d3, Andro LA 200-d3, Androtardyl-d3, Delatestryl-d3, Enarmone Depot-d3, Everone-d3, NSC 1759-d3, Primoteston-Depot-d3, Reposo TMD-d3, Testenate-d3, Testinon-d3, Testo-Enant-d3, Testosterone 17-Enanthate-d3, Testosterone Enanthate-d3, Testosterone Heptylate-d3, Testosterone Oenanthate-d3 |
| InChI Formula | InChI=1S/C26H40O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h17,20-23H,4-16H2,1-3H3/t20-,21-,22-,23-,25-,26-/m0/s1/i12D2,23D |
| SMILES | O=C1CC[C@@]2(C)C(CC[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC([2H])([2H])[C@]4([2H])OC(CCCCCC)=O)=C1 |
| IUPAC Name | (8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl-16,16,17-d3 heptanoate |
| Molecular Weight (g/mol) | 403.61 |
| Formula Weight | 403.32 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?