missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Antibody (3T16), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50332B
This item is not returnable.
View return policy
Description
Testosterone Monoclonal specifically detects Testosterone in Non-species specific samples. It is validated for ELISA.
Specifications
| Testosterone | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| Testosterone-3-(O-carboxymethyl)oxime coupled to bovine serum albumin | |
| 0.1 mL | |
| Cellular Signaling, Developmental Biology | |
| Non-species specific | |
| Purified |
| ELISA | |
| 3T16 | |
| ELISA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction