Learn More
Lenz Laborglasintrumente Test Tubes with NS Joint without Graduation
Product Code. 10593135
Click to view available options
Height (Metric):
100 mm
125 mm
Closure Type:
Hexagonal Stopper
Not Included
Unit Size:
Pack of 10
Product Code. 10593135
Brand: Lenz Laborglasintrumente 05700207
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With round bottom
  • With NS socket
  • Available without stopper
  • With PE stopper or with hexagonal hollow glass stopper
  • Made of DURAN™ tubing
Specifications
Closure Material Glass
Material Borosilicate Glass (3.3)
Height (Metric) 100 mm
Diameter (Metric) 16 mm
Closure Type Hexagonal Stopper