missing translation for 'onlineSavingsMsg'
Learn More

tert-Nitrobutane, 98+%

Product Code. p-7024619
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444157 5 g 5g
11454157 25 g 25g
11464157 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444157 Supplier Thermo Scientific Alfa Aesar Supplier No. A19608.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 594-70-7 | C4H9NO2 | 103.121 g/mol

It is used in the cyclic voltammetric study of tert-nitrobutane reduction in acetonitrile at mercury and platinum electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the cyclic voltammetric study of tert-nitrobutane reduction in acetonitrile at mercury and platinum electrodes.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-70-7
Molecular Formula C4H9NO2
Molecular Weight (g/mol) 103.121
MDL Number MFCD00007393
InChI Key AIMREYQYBFBEGQ-UHFFFAOYSA-N
Synonym propane, 2-methyl-2-nitro, trimethylnitromethane, 2-nitroisobutane, 2-nitro-2-methylpropane, tert-nitrobutane, 1,1-dimethyl-1-nitroethane, nitro-tert-butane, 2-methyl-2-nitro-propane, unii-9dkh5l679z, ccris 5044
PubChem CID 11672
IUPAC Name 2-methyl-2-nitropropane
SMILES CC(C)(C)[N+](=O)[O-]

Specifications

Melting Point 27°C to 31°C
Density 0.95
Boiling Point 126°C to 127°C
Flash Point 19°C (66°F)
Refractive Index 1.4
Quantity 5 g
UN Number UN1325
Beilstein 1744184
Solubility Information Insoluble in water.
Formula Weight 103.12
Percent Purity ≥98%
Chemical Name or Material tert-Nitrobutane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • tert-Nitrobutane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-methyl-2-nitropropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.