Learn More
tert-Butyldiphenylchlorosilane, TRC
Product Code. 30179517
Change view
Click to view available options
Quantity:
2 g
10 g
25 g
Unit Size:
10g
25g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179517 2 g 2g
30178970 10 g 10g
30179031 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179517 Supplier Toronto Research Chemicals Supplier No. TRCB6924152G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 58479-61-1
Molecular Formula C16H19ClSi
Molecular Weight (g/mol) 274.86
Synonym 1,1'-[Chloro(1,1-dimethylethyl)silylene]bisbenzene, Chloro(1,1-dimethylethyl)diphenylsilane, (1,1-Dimethylethyl)diphenylsilyl Chloride, (Chloro)(tert-butyl)diphenylsilane, Chloro(1,1-dimethylethyl)diphenylsilane, Chlorodiphenyl-tert-butylsilane, Diphenyl-tert-butylsilyl Chloride, NSC 617386, tert-Butylchlorodiphenylsilane, tert-Butyldiphenylsilyl Chloride, TBDPSCl, TBDPS-Cl
IUPAC Name tert-butyl-chloro-diphenylsilane
SMILES CC(C)(C)[Si](Cl)(c1ccccc1)c2ccccc2
Specifications
Quantity 2 g
InChI Formula InChI=1S/C16H19ClSi/c1-16(2,3)18(17,14-10-6-4-7-11-14)15-12-8-5-9-13-15/h4-13H,1-3H3
Formula Weight 274.0945
Chemical Name or Material tert-Butyldiphenylchlorosilane