missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride, 98%

Product Code. p-7055798
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475685 250 mg 250mg
15418946 1 g 1g
15428946 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15475685 Supplier Thermo Scientific Alfa Aesar Supplier No. H64468.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 77497-74-6 | C14H19NO2 | 233.311 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refrigerated.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77497-74-6
Molecular Formula C14H19NO2
Molecular Weight (g/mol) 233.311
MDL Number MFCD04114841
InChI Key KRSUDBIZXKOKGA-LBPRGKRZSA-N
Synonym s-tert-butyl 1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid t-butyl ester, tert-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert-butyl s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester, tic-obut, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic ac, tert-butyl s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, tert.-butyl 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylate, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid tert-butyl ester hydrochloride
PubChem CID 9209323
IUPAC Name tert-butyl (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate
SMILES CC(C)(C)OC(=O)C1CC2=CC=CC=C2CN1

Specifications

Quantity 250 mg
Beilstein 5744730
Solubility Information Soluble in water.
Formula Weight 269.77
Percent Purity 99%
Physical Form Solid
Chemical Name or Material tert-Butyl (S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.