Learn More
tert-Butyl Pitavastatin, TRC
Specifications
Specifications
| Chemical Name or Material | Pitavastatin tert-Butyl Ester |
| CAS | 586966-54-3 |
| Molecular Formula | C29 H32 F N O4 |
| Quantity | 100 mg |
| Synonym | (3R,5S,6E)-7-[2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoic acid 1,1-dimethylethyl ester, 1,1-Dimethylethyl (3R,5S,6E)-7-[2-cyclopropyl-4-(4-fluorophenyl)-3-quinolinyl]-3,5-dihydroxy-6-heptenoate, Pitavastatin tert-Butyl Ester |
| InChI Formula | InChI=1S/C29H32FNO4/c1-29(2,3)35-26(34)17-22(33)16-21(32)14-15-24-27(18-10-12-20(30)13-11-18)23-6-4-5-7-25(23)31-28(24)19-8-9-19/h4-7,10-15,19,21-22,32-33H,8-9,16-17H2,1-3H3/b15-14+/t21-,22-/m1/s1 |
| SMILES | CC(C)(C)OC(=O)C[C@H](O)C[C@H](O)\C=C\c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4 |
| IUPAC Name | tert-butyl (E,3R,5S)-7-[2-cyclopropyl-4-(4-fluorophenyl)quinolin-3-yl]-3,5-dihydroxyhept-6-enoate |
| Molecular Weight (g/mol) | 477.567 |
| Formula Weight | 477.232 |