Learn More
tert-Butyl Methyl Ether-d12, TRC
Product Code. 30452931
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30452931 5 mg 5mg
30455051 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30452931 Supplier Toronto Research Chemicals Supplier No. TRCT6933725MG
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl-tert-Butyl Ether D12
CAS 284487-64-5
Molecular Formula C5D12O
Quantity 5 mg
Synonym 1,1,1,3,3,3-Hexadeuterio-2-(trideuteriomethoxy)-2-(trideuteriomethyl)propane, tert-Butyl methyl ether-d12
InChI Formula InChI=1S/C5H12O/c1-5(2,3)6-4/h1-4H3/i1D3,2D3,3D3,4D3
SMILES [2H]C([2H])([2H])OC(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC Name 2-(methoxy-d3)-2-(methyl-d3)propane-1,1,1,3,3,3-d6
Molecular Weight (g/mol) 100.22
Formula Weight 100.16