missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 5-bromo-1H-indazole-1-carboxylate, ≥90%, Thermo Scientific™

Product Code. p-3174939
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10295844 250 mg Amber Glass Bottle 250mg
10552064 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10295844 Supplier Thermo Scientific Maybridge Supplier No. CC53210CB

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 651780-02-8
Molecular Formula C12H13BrN2O2
Molecular Weight (g/mol) 297.152
MDL Number MFCD08435913
InChI Key CAQGDWLRIQCXSP-UHFFFAOYSA-N
PubChem CID 22832025
IUPAC Name tert-butyl 5-bromoindazole-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C2=C(C=C(C=C2)Br)C=N1

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym tert-butyl 5-bromo-1h-indazole-1-carboxylate, 1-boc-5-bromo-indazole, n-boc-5-bromo-1h-indazole, n-boc-5-bromoindazole, 1-boc-5-bromo-1h-indazole, n-boc-5-bromo indazole, 1h-indazole-1-carboxylic acid, 5-bromo-, 1,1-dimethylethyl ester, tert-butyl5-bromo-1h-indazole-1-carboxylate, 1-tert-butoxycarbonyl-5-bromoindazole, 5-bromoindazole-1-carboxylic acid tert-butyl ester
Formula Weight 297.15
Percent Purity 90%
Chemical Name or Material Tert-butyl 5-bromo-1H-indazole-1-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.