missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-[(trifluoromethyl)sulfonyloxy]-5,6-dihydropyridine-1(2h)-carboxylate, 90%, Thermo Scientific™

Product Code. 10105663
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10105663 5 g Amber Glass Bottle 5g
10239333 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10105663 Supplier Thermo Scientific Maybridge Supplier No. MO07986DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 138647-49-1
Molecular Formula C11H16F3NO5S
Molecular Weight (g/mol) 331.31
MDL Number MFCD09997858
InChI Key WUBVEMGCQRSBBT-UHFFFAOYSA-N
Synonym tert-butyl 4-trifluoromethyl sulfonyl oxy-5,6-dihydropyridine-1 2h-carboxylate, 1-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridin-4-yl trifluoromethanesulfonate, n-boc-4-trifluoromethanesulfonyloxy-3,6-dihydro-2h-pyridine, 1-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridin-4-yl-trifluoromethanesulfonate, tert-butyl 4-trifluoromethylsulfonyloxy-3,6-dihydro-2h-pyridine-1-carboxylate, tert-butyl 4-trifluoromethylsulfonyloxy-5,6-dihydropyridine-1 2h-carboxylate, 4-trifluoromethanesulfonyloxy-3,6-dihydro-2h-pyridine-1-carboxylic acid tert-butyl ester, 1 2h-pyridinecarboxylic acid, 3,6-dihydro-4-trifluoromethyl sulfonyl oxy-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-1,2,3,6-tetrahydropyridin-4-yltrifluoromethanesulfonate, tert-butyl 4-trifluoromethane sulfonyloxy-1,2,3,6-tetrahydropyridine-1-carboxylate
PubChem CID 10991161
IUPAC Name tert-butyl 4-(trifluoromethylsulfonyloxy)-3,6-dihydro-2H-pyridine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(OS(=O)(=O)C(F)(F)F)=CC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 331.31
Percent Purity 97%
Chemical Name or Material Tert-butyl 4-[(trifluoromethyl)sulfonyloxy]-5, 6-dihydropyridine-1(2h)-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.